Learn More
Uvex 95790 Insole For Uvex 1 Sport - Width 14
Comfortable climatic insole with antistatic properties for Uvex 1 Sport
Brand: Uvex 9579043
136.67 NOK valid until 2023-05-26
Use promo code "20831" to get your promotional price.
Additional Details : Weight : 0.08200kg
Description
- Full-surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No. 9579.0
- Sizes: 35 to 52
Specifications
Black | |
Microvelour | |
No | |
Wide width | |
No |
Uvex 1 Sport | |
43 | |
Unisex | |
Yes |