missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Phenylcarbazole-3-boronic acid, 98%

Brand:  Alfa Aesar™ H64059.MD

Additional Details : CAS Number : 854952-58-2 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15431089

  • 253.98 NOK / 250mg


Estimated Shipment
11-08-2020
Add to basket

Description

Description

Applications
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

854952-58-2
287.125
JWJQEUDGBZMPAX-UHFFFAOYSA-N
51358450
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)C4=CC=CC=C4)(O)O
C18H14BNO2
MFCD12196936
9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int
(9-phenylcarbazol-3-yl)boronic acid

Specifications

9-Phenylcarbazole-3-boronic acid
854952-58-2
MFCD12196936
9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int
JWJQEUDGBZMPAX-UHFFFAOYSA-N
(9-phenylcarbazol-3-yl)boronic acid
51358450
Solid
250mg
C18H14BNO2
10566519
Slightly soluble in water.
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)C4=CC=CC=C4)(O)O
287.125
287.13
98%
Videos
SDS Documents