missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 9-Isopropylcarbazole, 97%
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 118°C to 122°C |
Quantity | 5 g |
Formula Weight | 209.29 |
Percent Purity | 97% |
Chemical Name or Material | 9-Isopropylcarbazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'