missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7034280
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
569343-09-5
C21H25BO2
320.239
MFCD08704229
DAZFRJAIIUPRQZ-UHFFFAOYSA-N
2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2-boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl, 2-9,9-dimethylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fl-be, amtb252, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9,9-dimethyl-9h-fluorene
45358694
2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(C)C
This item is not returnable. View return policy
569343-09-5
C21H25BO2
320.239
MFCD08704229
DAZFRJAIIUPRQZ-UHFFFAOYSA-N
2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2-boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl, 2-9,9-dimethylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fl-be, amtb252, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9,9-dimethyl-9h-fluorene
45358694
2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(C)C

It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 320.24
Percent Purity 95%
Chemical Name or Material 9,9-Dimethylfluorene-2-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'