missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7052698
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
317802-08-7
C35H52B2O4
558.42
MFCD03701607
KAYXDWIILRESPY-UHFFFAOYSA-N
9,9-dioctylfluorene-2,7-diboronic acid bis 1,3-propanediol ester, 1,3,2-dioxaborinane, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethylborate, 9 9-dioctylfluorene-2 7-bis trimethylbo, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctylfluoren-2-yl-1,3,2-dioxaborinane, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctyl-fluoren-2-yl-1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethyleneborate, 2,2-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane
4378560
2-[7-(1,3,2-dioxaborinan-2-yl)-9,9-dioctylfluoren-2-yl]-1,3,2-dioxaborinane
CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OCCCO2)C2=C1C=C(C=C2)B1OCCCO1
This item is not returnable. View return policy
317802-08-7
C35H52B2O4
558.42
MFCD03701607
KAYXDWIILRESPY-UHFFFAOYSA-N
9,9-dioctylfluorene-2,7-diboronic acid bis 1,3-propanediol ester, 1,3,2-dioxaborinane, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethylborate, 9 9-dioctylfluorene-2 7-bis trimethylbo, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctylfluoren-2-yl-1,3,2-dioxaborinane, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctyl-fluoren-2-yl-1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethyleneborate, 2,2-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane
4378560
2-[7-(1,3,2-dioxaborinan-2-yl)-9,9-dioctylfluoren-2-yl]-1,3,2-dioxaborinane
CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OCCCO2)C2=C1C=C(C=C2)B1OCCCO1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 558.42
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'