missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-(Trifluoromethyl)quinoline, 97%

Brand:  Alfa Aesar™ H27771.03

Additional Details : CAS Number : 325-14-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11392939

  • 1737.00 NOK / 1g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

325-14-4
197.16
CMMSEFHVUYEEDY-UHFFFAOYSA-N
459614
C1=CC2=C(C=C(C=C2)C(F)(F)F)N=C1
C10H6F3N
MFCD00833760
7-trifluoromethyl quinoline, 7-cf3 quinoline, 7-trrfluoromethylquinoline, pubchem7218, 7-trifluoromethyl-quinoline, acmc-20a40f, quinoline,7-trifluoromethyl, quinoline, 7-trifluoromethyl
7-(trifluoromethyl)quinoline

Specifications

7-(Trifluoromethyl)quinoline
1g
C10H6F3N
7-trifluoromethyl quinoline, 7-cf3 quinoline, 7-trrfluoromethylquinoline, pubchem7218, 7-trifluoromethyl-quinoline, acmc-20a40f, quinoline,7-trifluoromethyl, quinoline, 7-trifluoromethyl
C1=CC2=C(C=C(C=C2)C(F)(F)F)N=C1
197.16
197.16
65°C to 66°C
325-14-4
MFCD00833760
CMMSEFHVUYEEDY-UHFFFAOYSA-N
7-(trifluoromethyl)quinoline
459614
97%
Videos
SDS Documents