missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine, 95%, Thermo Scientific™

Catalog Number p-2703504
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
154264-95-6
C9H10BrNO
228.09
MFCD02681913
MQMFOFZKZBLSAB-UHFFFAOYSA-N
7-bromo-4-methyl-3,4-dihydro-2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl-2h-1,4-benzoxazine, 2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-bromo-4-methyl-2h,3h-benzo e 1,4-oxazine, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine
2776405
7-bromo-4-methyl-2,3-dihydro-1,4-benzoxazine
CN1CCOC2=C1C=CC(Br)=C2
This item is not returnable. View return policy
154264-95-6
C9H10BrNO
228.09
MFCD02681913
MQMFOFZKZBLSAB-UHFFFAOYSA-N
7-bromo-4-methyl-3,4-dihydro-2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl-2h-1,4-benzoxazine, 2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-bromo-4-methyl-2h,3h-benzo e 1,4-oxazine, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine
2776405
7-bromo-4-methyl-2,3-dihydro-1,4-benzoxazine
CN1CCOC2=C1C=CC(Br)=C2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 228.09
Percent Purity 95%
Chemical Name or Material 7-Bromo-4-methyl-3, 4-dihydro-2H-1, 4-benzoxazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'