missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Nitroindole-3-carboxaldehyde, 95%

Brand:  Alfa Aesar™ H63674.MD

Additional Details : CAS Number : 10553-13-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15430489

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

10553-13-6
190.158
KYFZSGBVGJNEPN-UHFFFAOYSA-N
5210076
C1=CC2=C(C=C1[N+](=O)[O-])NC=C2C=O
C9H6N2O3
MFCD06656872
6-nitro-1h-indole-3-carboxaldehyde, 6-nitroindole-3-carboxaldehyde, 6-nitro-3-indolecarbaldehyde, 6-nitroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde,6-nitro, 6-nitro-1h-indole-3-carbaldehyde
6-nitro-1H-indole-3-carbaldehyde

Specifications

6-Nitroindole-3-carboxaldehyde
10553-13-6
MFCD06656872
6-nitro-1h-indole-3-carboxaldehyde, 6-nitroindole-3-carboxaldehyde, 6-nitro-3-indolecarbaldehyde, 6-nitroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde,6-nitro, 6-nitro-1h-indole-3-carbaldehyde
C1=CC2=C(C=C1[N+](=O)[O-])NC=C2C=O
190.158
190.16
95%
250mg
C9H6N2O3
165207
KYFZSGBVGJNEPN-UHFFFAOYSA-N
6-nitro-1H-indole-3-carbaldehyde
5210076
Solid
Videos
SDS Documents