Learn More
6-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
Catalog Number p-7032441
Click to view available options
Quantity:
250 mg
1 g
28226-22-4
C8H8N2O3
180.163
MFCD07698591
GZAJZBARYACGSO-UHFFFAOYSA-N
6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-6-nitro, 2h-1,4-benzoxazine, 3,4-dihydro-6-nitro, 6-nitro-2,3-dihydrobenz-1,4-oxazine, 6-nitro-2h,3h,4h-benzo e 1,4-oxazine, 6-nitro-3,4-dihydro-2h-1,4 benzoxazine, 6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester
7062225
6-nitro-3,4-dihydro-2H-1,4-benzoxazine
C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]
28226-22-4
C8H8N2O3
180.163
MFCD07698591
GZAJZBARYACGSO-UHFFFAOYSA-N
6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-6-nitro, 2h-1,4-benzoxazine, 3,4-dihydro-6-nitro, 6-nitro-2,3-dihydrobenz-1,4-oxazine, 6-nitro-2h,3h,4h-benzo e 1,4-oxazine, 6-nitro-3,4-dihydro-2h-1,4 benzoxazine, 6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester
7062225
6-nitro-3,4-dihydro-2H-1,4-benzoxazine
C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 117°C to 119°C
Quantity 250 mg
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material 6-Nitro-3,4-dihydro-2H-1,4-benzoxazine