missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole-3-carboxaldehyde, 98%

Product Code. p-7053906
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15418228

Brand: Thermo Scientific Alfa Aesar H61665.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2795-41-7 | C9H6FNO | 163.15 g/mol

6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2795-41-7
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00069703
InChI Key CWCYUOSLRVAKQZ-UHFFFAOYSA-N
Synonym 6-fluoroindole-3-carboxaldehyde, 6-fluoroindole-3-carbaldehyde, 6-fluoroindole-3-aldehyde, 6-fluoro-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 6-fluoro, 1h-6-fluoroindole-3-carboxaldehyde, pubchem7269, ksc495g4p, 6-fluoro-3-formyl-1h-indole, 6-fluorooindole-3-carboxaldehyde
PubChem CID 262903
IUPAC Name 6-fluoro-1H-indole-3-carbaldehyde
SMILES FC1=CC=C2C(NC=C2C=O)=C1

Specifications

Color Yellow
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 163.15
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 6-Fluoroindole-3-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.