missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,3-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7032217
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
887584-84-1
C7H3ClF2O2
192.546
MFCD07368094
MJWXJSUYTVBLJU-UHFFFAOYSA-N
2,3-difluoro-6-chlorobenzoic acid
29922009
6-chloro-2,3-difluorobenzoic acid
C1=CC(=C(C(=C1F)F)C(=O)O)Cl
This item is not returnable. View return policy
887584-84-1
C7H3ClF2O2
192.546
MFCD07368094
MJWXJSUYTVBLJU-UHFFFAOYSA-N
2,3-difluoro-6-chlorobenzoic acid
29922009
6-chloro-2,3-difluorobenzoic acid
C1=CC(=C(C(=C1F)F)C(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 192.55
Percent Purity 97%
Chemical Name or Material 6-Chloro-2,3-difluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'