missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15430689
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
16732-65-3
C9H6BrNO2
240.056
MFCD02664469
SVBVYRYROZWKNJ-UHFFFAOYSA-N
6-bromoindole-2-carboxylic acid, 6-bromo-2-indolecarboxylic acid, 1h-indole-2-carboxylic acid, 6-bromo, 6-bromoindole-2-carboxylicacid, zlchem 475, pubchem1664, acmc-1bqkh, intermediates-zcf02255, evoblocks eb21020, timtec-bb sbb014280
4011696
6-bromo-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1Br)NC(=C2)C(=O)O
This item is not returnable. View return policy
16732-65-3
C9H6BrNO2
240.056
MFCD02664469
SVBVYRYROZWKNJ-UHFFFAOYSA-N
6-bromoindole-2-carboxylic acid, 6-bromo-2-indolecarboxylic acid, 1h-indole-2-carboxylic acid, 6-bromo, 6-bromoindole-2-carboxylicacid, zlchem 475, pubchem1664, acmc-1bqkh, intermediates-zcf02255, evoblocks eb21020, timtec-bb sbb014280
4011696
6-bromo-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1Br)NC(=C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 240.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromoindole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'