missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-8-iodoimidazo[1,2-a]pyridine, 95%, Thermo Scientific™

Catalog Number p-7052597
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1364917-14-5
C7H4BrIN2
322.931
MFCD21609484
OKEMQQRZPHOOGS-UHFFFAOYSA-N
71299170
6-bromo-8-iodoimidazo[1,2-a]pyridine
C1=CN2C=C(C=C(C2=N1)I)Br
This item is not returnable. View return policy
1364917-14-5
C7H4BrIN2
322.931
MFCD21609484
OKEMQQRZPHOOGS-UHFFFAOYSA-N
71299170
6-bromo-8-iodoimidazo[1,2-a]pyridine
C1=CN2C=C(C=C(C2=N1)I)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 150°C
Color Yellow
Quantity 1 g
Sensitivity Light sensitive
Synonym 6-bromo-8-iodoimidazo 1,2-a pyridine, 6-bromo-8-iodo-imidazo 1,2-a pyridine, 6-bromo-8-iodoimidazo 1,2-a pyridine,, imidazo 1,2-a pyridine,6-bromo-8-iodo, imidazo 1,2-a pyridine, 6-bromo-8-iodo
Formula Weight 322.92
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6-Bromo-8-iodoimidazo[1,2-a]pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'