missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-5-methylpyridine-3-boronic acid, 95%, Thermo Scientific™

Catalog Number p-7029725
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1003043-34-2
C6H7BBrNO2
215.841
MFCD08274468
IPCXBKSYMWZLBR-UHFFFAOYSA-N
2-bromo-3-methylpyridine-5-boronic acid, 6-bromo-5-methylpyridine-3-boronic acid, 6-bromo-5-methylpyridin-3-yl boronic acid, 2-bromo-3-methyl-5-pyridineboronic acid, 5-borono-2-bromo-3-picoline, 6-bromo-5-methylpyridin-3-yl-3-boronic acid, 2-bromo-3-picoline-5-boronic acid, 6-bromo-5-methyl-3-pyridyl boronic acid, pubchem5081, acmc-2097oc
17750219
(6-bromo-5-methylpyridin-3-yl)boronic acid
B(C1=CC(=C(N=C1)Br)C)(O)O
This item is not returnable. View return policy
1003043-34-2
C6H7BBrNO2
215.841
MFCD08274468
IPCXBKSYMWZLBR-UHFFFAOYSA-N
2-bromo-3-methylpyridine-5-boronic acid, 6-bromo-5-methylpyridine-3-boronic acid, 6-bromo-5-methylpyridin-3-yl boronic acid, 2-bromo-3-methyl-5-pyridineboronic acid, 5-borono-2-bromo-3-picoline, 6-bromo-5-methylpyridin-3-yl-3-boronic acid, 2-bromo-3-picoline-5-boronic acid, 6-bromo-5-methyl-3-pyridyl boronic acid, pubchem5081, acmc-2097oc
17750219
(6-bromo-5-methylpyridin-3-yl)boronic acid
B(C1=CC(=C(N=C1)Br)C)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 215.84
Percent Purity 95%
Chemical Name or Material 6-Bromo-5-methylpyridine-3-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'