Learn More
6-Aminoquinoline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.
Solubility
Soluble in methanol, chloroform, ethanol, and benzene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.
Solubility
Soluble in methanol, chloroform, ethanol, and benzene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications
Melting Point | 115°C to 119°C |
Boiling Point | 146°C (0.3 mmHg) |
Odor | Odorless |
Quantity | 1 g |
Beilstein | 113320 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in methanol,chloroform,ethanol,and benzene. Insoluble in water. |
Formula Weight | 144.18 |
Percent Purity | 98% |
Chemical Name or Material | 6-Aminoquinoline |