missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Amino-2,3-difluorophenol, 95%

Brand:  Alfa Aesar™ H63249.06

Additional Details : CAS Number : 115551-33-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429848

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

115551-33-2
145.109
KRURHZHJEDNBCM-UHFFFAOYSA-N
2778441
C1=CC(=C(C(=C1N)O)F)F
C6H5F2NO
MFCD03094310
3,4-difluoro-2-hydroxyaniline, 6-amino-2,3-difluoro-phenol, phenol, 6-amino-2,3-difluoro, phenol,6-amino-2,3-difluoro, pubchem3544, acmc-2099qh, 2-amino-5,6-difluorophenol, 2,3-difluoro-6-aminophenol, 6-amino-2,3-diflourophenol, 6-amino-2,3 difluoro-phenol
6-amino-2,3-difluorophenol

Specifications

6-Amino-2,3-difluorophenol
115551-33-2
MFCD03094310
KRURHZHJEDNBCM-UHFFFAOYSA-N
6-amino-2,3-difluorophenol
2778441
Solid
5g
C6H5F2NO
3,4-difluoro-2-hydroxyaniline, 6-amino-2,3-difluoro-phenol, phenol, 6-amino-2,3-difluoro, phenol,6-amino-2,3-difluoro, pubchem3544, acmc-2099qh, 2-amino-5,6-difluorophenol, 2,3-difluoro-6-aminophenol, 6-amino-2,3-diflourophenol, 6-amino-2,3 difluoro-phenol
C1=CC(=C(C(=C1N)O)F)F
145.109
145.11
95%
Videos
SDS Documents