missing translation for 'onlineSavingsMsg'
Learn More

6,7,8,9,10,11-Hexahydrocyclooct[b]indole, 97%, Thermo Scientific™

Catalog Number p-7024320
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
22793-63-1
C14H17N
199.30
MFCD00086346
TYLZSGIRJRUUSQ-UHFFFAOYSA-N
6,7,8,9,10,11-hexahydro-5h-cycloocta b indole, 5h,6h,7h,8h,9h,10h,11h-cycloocta b indole, 5h-cyclooct b indole,6,7,8,9,10,11-hexahydro, acmc-20am0o, maybridge1_006281, tylzsgirjruusq-uhfffaoysa, 6,7,8,9,10,11-hexahydro-5h-cyclooct b indole, 6,7,8,9,10,11-hexahydrocycloocta 1,2-b indole, 6,7,8,9,10,11-hexahydrocycloocta 2,1-b indole
272686
5H,6H,7H,8H,9H,10H,11H-cycloocta[b]indole
C1CCCC2=C(CC1)NC1=CC=CC=C21
This item is not returnable. View return policy
22793-63-1
C14H17N
199.30
MFCD00086346
TYLZSGIRJRUUSQ-UHFFFAOYSA-N
6,7,8,9,10,11-hexahydro-5h-cycloocta b indole, 5h,6h,7h,8h,9h,10h,11h-cycloocta b indole, 5h-cyclooct b indole,6,7,8,9,10,11-hexahydro, acmc-20am0o, maybridge1_006281, tylzsgirjruusq-uhfffaoysa, 6,7,8,9,10,11-hexahydro-5h-cyclooct b indole, 6,7,8,9,10,11-hexahydrocycloocta 1,2-b indole, 6,7,8,9,10,11-hexahydrocycloocta 2,1-b indole
272686
5H,6H,7H,8H,9H,10H,11H-cycloocta[b]indole
C1CCCC2=C(CC1)NC1=CC=CC=C21

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 72°C to 76°C
Quantity 1 g
Formula Weight 199.3
Percent Purity 97%
Chemical Name or Material 6,7,8,9,10,11-Hexahydrocyclooct[b]indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'