Learn More
5-Thio-D-glucose, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Description
5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis. It is also used to control/reduce the rate of glycolysis in vivo.
Solubility
Miscible with water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis. It is also used to control/reduce the rate of glycolysis in vivo.
Solubility
Miscible with water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 135°C to 138°C |
Color | White |
Quantity | 5 mg |
Beilstein | 1865193 |
Merck Index | 14,9334 |
Formula Weight | 196.22 |
Percent Purity | ≥97% |
Physical Form | Powder |
Chemical Name or Material | 5-Thio-D-glucose |