missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (606112) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5795JF585
This item is not returnable.
View return policy
Description
5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Equine,Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| 5'-Nucleotidase/CD73 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
| Chinese hamster ovary cell line CHO-derived recombinant human 5'-Nucleotidase/CD73, Trp27-Ile511, Accession # P21589 | |
| RUO | |
| Primary | |
| Equine, Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 606112 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 4907 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction