missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-3-(1-pyrrolidinylmethyl)indole , 95%

Brand:  Alfa Aesar™ H63372.MD

Additional Details : CAS Number : 3414-68-4 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15430039

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

3414-68-4
245.282
CBLKOOHEMGHOAA-UHFFFAOYSA-N
24839583
C1CCN(C1)CC2=CNC3=C2C=C(C=C3)[N+](=O)[O-]
C13H15N3O2
MFCD01718616
5-nitro-3-pyrrolidin-1-ylmethyl-1h-indole, 5-nitro-3-1-pyrrolidinylmethyl-1h-indole, 5-nitro-3-pyrrolidin-1-yl methyl-1h-indole
5-nitro-3-(pyrrolidin-1-ylmethyl)-1H-indole

Specifications

5-Nitro-3-(1-pyrrolidinylmethyl)indole
3414-68-4
MFCD01718616
5-nitro-3-pyrrolidin-1-ylmethyl-1h-indole, 5-nitro-3-1-pyrrolidinylmethyl-1h-indole, 5-nitro-3-pyrrolidin-1-yl methyl-1h-indole
C1CCN(C1)CC2=CNC3=C2C=C(C=C3)[N+](=O)[O-]
245.282
245.28
95%
250mg
C13H15N3O2
421075
CBLKOOHEMGHOAA-UHFFFAOYSA-N
5-nitro-3-(pyrrolidin-1-ylmethyl)-1H-indole
24839583
Solid
Videos
SDS Documents