Learn More
Fisherbrand™ 5 Minute Marking, Transparent Pocket Timer
Easy-to-read, and use—no keys, no batteries
Brand: Fisherbrand™
Additional Details : Weight : 0.09070kg
Description
- Count down
- Key-chain
- Lanyard
- Loud five-second bell rings at zero
Specifications
Countdown Timer | |
Audible 5 second alarm | |
1 min. | |
None | |
0.2 lb. |
1 to 60 min. | |
Not Tested | |
1 min. | |
Blue | |
0.09 kg |