Learn More
Alfa Aesar™ 5-Methylthiazole, 97%
Brand: Alfa Aesar™ L15879.03
Additional Details : CAS Number : 3581-89-3 Weight : 0.00100kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Quantity | 1g |
---|
Description
Applications5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
3581-89-3 | |
99.151 | |
RLYUNPNLXMSXAX-UHFFFAOYSA-N | |
137980 | |
CC1=CN=CS1 |
C4H5NS | |
MFCD00005335 | |
5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d | |
5-methyl-1,3-thiazole |
Specifications
5-Methylthiazole | |
42°C (107°F) | |
1.528 | |
1g | |
C4H5NS | |
UN1993 | |
5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d | |
RLYUNPNLXMSXAX-UHFFFAOYSA-N | |
5-methyl-1,3-thiazole | |
137980 | |
97% |
1.12 | |
Stench | |
141°C to 142°C | |
3581-89-3 | |
MFCD00005335 | |
104674 | |
Insoluble in water. | |
CC1=CN=CS1 | |
99.151 | |
99.16 |