missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7046701
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3581-89-3
C4H5NS
99.151
MFCD00005335
RLYUNPNLXMSXAX-UHFFFAOYSA-N
5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
137980
5-methyl-1,3-thiazole
CC1=CN=CS1
This item is not returnable. View return policy
3581-89-3
C4H5NS
99.151
MFCD00005335
RLYUNPNLXMSXAX-UHFFFAOYSA-N
5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
137980
5-methyl-1,3-thiazole
CC1=CN=CS1

5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.12
Boiling Point 141°C to 142°C
Flash Point 42°C (107°F)
Odor Stench
Refractive Index 1.528
Quantity 1 g
UN Number UN1993
Beilstein 104674
Solubility Information Insoluble in water.
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material 5-Methylthiazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'