missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylisoxazole-3-carboxylic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
It is a reactant for Click/aza-Michael or Click/oligomeric alkyl carbodiimide esterification reactions, preparation of aminopyrazole amide derivatives as Raf kinase inhibitors in melanoma cells and preparation of trifluoromethoxyphenyl(thiazolyl)pyrroles and other heterocycle-bearing polyfunctionalized pyrroles via chain heterocyclization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a reactant for Click/aza-Michael or Click/oligomeric alkyl carbodiimide esterification reactions, preparation of aminopyrazole amide derivatives as Raf kinase inhibitors in melanoma cells and preparation of trifluoromethoxyphenyl(thiazolyl)pyrroles and other heterocycle-bearing polyfunctionalized pyrroles via chain heterocyclization.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
It is a reactant for Click/aza-Michael or Click/oligomeric alkyl carbodiimide esterification reactions, preparation of aminopyrazole amide derivatives as Raf kinase inhibitors in melanoma cells and preparation of trifluoromethoxyphenyl(thiazolyl)pyrroles and other heterocycle-bearing polyfunctionalized pyrroles via chain heterocyclization.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
Melting Point | 171°C to 173°C |
Quantity | 5 g |
Beilstein | 114094 |
Formula Weight | 127.1 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-Methylisoxazole-3-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'