missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexanone, 99%, Thermo Scientific Chemicals

Catalog Number p-7047093
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
110-12-3
C7H14O
114.188
MFCD00008950
FFWSICBKRCICMR-UHFFFAOYSA-N
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
8034
5-methylhexan-2-one
CC(C)CCC(=O)C
This item is not returnable. View return policy
110-12-3
C7H14O
114.188
MFCD00008950
FFWSICBKRCICMR-UHFFFAOYSA-N
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
8034
5-methylhexan-2-one
CC(C)CCC(=O)C

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (5.4 g/L)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -74°C
Density 0.812
Boiling Point 144°C to 145°C
Flash Point 41°C (105°F)
Refractive Index 1.4075
Quantity 100 mL
UN Number UN2302
Beilstein 506163
Solubility Information Soluble in water. (5.4g/L)
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hexanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'