missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methoxy-3-(1-pyrrolidinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63433.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430129

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

885526-18-1
230.31
VIWIIMCQNDZROC-UHFFFAOYSA-N
69340661
C14H18N2O
MFCD28098705
5-methoxy-3-pyrrolidin-1-ylmethyl-1h-indole, 5-methoxy-3-1-pyrrolidinylmethyl indole, 5-methoxy-3-pyrrolidin-1-yl methyl-1h-indole

Specifications

5-Methoxy-3-(1-pyrrolidinylmethyl)indole
885526-18-1
MFCD28098705
VIWIIMCQNDZROC-UHFFFAOYSA-N
69340661
Solid
5g
C14H18N2O
5-methoxy-3-pyrrolidin-1-ylmethyl-1h-indole, 5-methoxy-3-1-pyrrolidinylmethyl indole, 5-methoxy-3-pyrrolidin-1-yl methyl-1h-indole
230.31
230.31
95%
Videos
SDS Documents