missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyflavone, 97%, Thermo Scientific Chemicals

Catalog Number p-7046401
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
491-78-1
C15H10O3
238.24
MFCD00016944
IYBLVRRCNVHZQJ-UHFFFAOYSA-N
5-hydroxyflavone, primuletin, 5-hydroxy-2-phenyl-4h-chromen-4-one, 5-hydroxy-2-phenyl-chromen-4-one, chembl16807, 4h-1-benzopyran-4-one,5-hydroxy-2-phenyl, primuliten, 5-hydroxy-flavone, 5-hydroxy-2-phenyl-4-benzopyrone, 5-hydroxy-2-phenyl-4h-chromen-4-on
68112
5-hydroxy-2-phenylchromen-4-one
OC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
491-78-1
C15H10O3
238.24
MFCD00016944
IYBLVRRCNVHZQJ-UHFFFAOYSA-N
5-hydroxyflavone, primuletin, 5-hydroxy-2-phenyl-4h-chromen-4-one, 5-hydroxy-2-phenyl-chromen-4-one, chembl16807, 4h-1-benzopyran-4-one,5-hydroxy-2-phenyl, primuliten, 5-hydroxy-flavone, 5-hydroxy-2-phenyl-4-benzopyrone, 5-hydroxy-2-phenyl-4h-chromen-4-on
68112
5-hydroxy-2-phenylchromen-4-one
OC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 160°C
Quantity 100 mg
Beilstein 194429
Formula Weight 238.25
Percent Purity 97%
Chemical Name or Material 5-Hydroxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'