missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexynoic acid, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Density | 0.99 |
Boiling Point | 73°C to 74°C (0.2 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.4495 |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 1743192 |
Solubility Information | Miscible with water. |
Formula Weight | 112.13 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'