missing translation for 'onlineSavingsMsg'
Learn More

5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog Number p-7059104
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
16219-75-3
C9H12
120.195
MFCD00167576
OJOWICOBYCXEKR-WHZSQGQPSA-N
5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component
12456033
(1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
CC=C1CC2CC1C=C2
This item is not returnable. View return policy
16219-75-3
C9H12
120.195
MFCD00167576
OJOWICOBYCXEKR-WHZSQGQPSA-N
5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component
12456033
(1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
CC=C1CC2CC1C=C2

5-Ethylidene-2-norbornene is used as a self healing agent for polymeric composites. It is also used to reinforce multiwalled carbon nanotubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.89 g/mL
Boiling Point 146°C
Flash Point 38°C (101°F)
Quantity 25 mL
UN Number UN3295
Solubility Information Soluble in water (0.01% at 20C).
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Ethylidene-2-norbornene, endo + exo, Stabilized with 100ppm 4-tert-butylcatechol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'