Learn More
Alfa Aesar™ 5-Dimethylamino-2-methyl-3-pentyn-2-ol, 98%
Brand: Alfa Aesar™ L12256.06
Additional Details : CAS Number : 25400-83-3 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
25400-83-3 | |
141.214 | |
VBARYRPUABJYGW-UHFFFAOYSA-N | |
180775 | |
CC(C)(C#CCN(C)C)O |
C8H15NO | |
MFCD00041575 | |
5-dimethylamino-2-methyl-3-pentyn-2-ol, 5-dimethylamino-2-methylpent-3-yn-2-ol, 3-pentyn-2-ol, 5-dimethylamino-2-methyl, acmc-1cdlj, 5-dimethylamino-2-methyl-pent-3-yn-2-ol, 3-pentyn-2-ol,5-dimethylamino-2-methyl | |
5-(dimethylamino)-2-methylpent-3-yn-2-ol |
Specifications
5-Dimethylamino-2-methyl-3-pentyn-2-ol | |
91°C to 92°C (1mmHg) | |
25400-83-3 | |
MFCD00041575 | |
5-dimethylamino-2-methyl-3-pentyn-2-ol, 5-dimethylamino-2-methylpent-3-yn-2-ol, 3-pentyn-2-ol, 5-dimethylamino-2-methyl, acmc-1cdlj, 5-dimethylamino-2-methyl-pent-3-yn-2-ol, 3-pentyn-2-ol,5-dimethylamino-2-methyl | |
CC(C)(C#CCN(C)C)O | |
141.214 | |
141.21 |
1.4561 | |
5g | |
C8H15NO | |
1750347 | |
VBARYRPUABJYGW-UHFFFAOYSA-N | |
5-(dimethylamino)-2-methylpent-3-yn-2-ol | |
180775 | |
98% |