Learn More
5-Cyano-2-methylpyridine, 97%, Acros Organics
Brand:  Acros Organics 452420050
Additional Details : CAS Number : 3222-48-8 Weight : 0.01000kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity 5g
 View more versions of this product
Product Code. 15269816
  • 1715.46 NOK / 5g
Estimated Shipment date
from Supplier 16-06-2020
Add to basket
Specifications
Chemical Identifiers
3222-48-8
118.14
5-cyano-2-picoline, 5-cyano-2-methylpyridine, 6-methylnicotinonitrile, 6-methyl-3-pyridinecarbonitrile, 3-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-methyl, pubchem6695, 2-methyl-5-cyanopyridine, 6-methylnicotinonitrile #, acmc-209z1l
6-methylpyridine-3-carbonitrile
C7H6N2
PBLOYQAQGYUPCM-UHFFFAOYSA-N
281995
CC1=NC=C(C=C1)C#N
Specifications
5-Cyano-2-methylpyridine
5g
96% min. (GC)
5-cyano-2-picoline, 5-cyano-2-methylpyridine, 6-methylnicotinonitrile, 6-methyl-3-pyridinecarbonitrile, 3-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-methyl, pubchem6695, 2-methyl-5-cyanopyridine, 6-methylnicotinonitrile #, acmc-209z1l
CC1=NC=C(C=C1)C#N
118.14
118.14
81°C to 86°C
3222-48-8
C7H6N2
PBLOYQAQGYUPCM-UHFFFAOYSA-N
6-methylpyridine-3-carbonitrile
281995
97%
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 5-Cyano-2-methylpyridine, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Documents