missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(2-fluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64235.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15431339

  • 3570.00 NOK / 1g


Estimated Shipment date
from Supplier 15-10-2019
Add to basket

Specifications

Chemical Identifiers

MFCD09259577

Specifications

1g
MFCD09259577
Solid
5-Chloro-2-(2-fluorophenyl)benzimidazole
246.67
95%
Videos
SDS Documents