Learn More
5-Bromotryptamine hydrochloride, 98%, Thermo Scientific™
Catalog Number p-7047547
Click to view available options
Quantity:
250 mg
1 g
81868-12-4
C10H12BrClN2
275.574
MFCD00152022
QUPXCOYSKSUJPX-UHFFFAOYSA-N
2-5-bromo-1h-indol-3-yl ethanamine hydrochloride, 5-bromotryptamine hydrochloride, 2-5-bromo-1h-indol-3-yl-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-bromo-, monohydrochloride, 5-bromotryptamine hcl, 2-5-bromo-1h-indol-3-yl ethanaminehydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine hydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine-hydrogen chloride 1/1
13241175
2-(5-bromo-1H-indol-3-yl)ethanamine;hydrochloride
C1=CC2=C(C=C1Br)C(=CN2)CCN.Cl
81868-12-4
C10H12BrClN2
275.574
MFCD00152022
QUPXCOYSKSUJPX-UHFFFAOYSA-N
2-5-bromo-1h-indol-3-yl ethanamine hydrochloride, 5-bromotryptamine hydrochloride, 2-5-bromo-1h-indol-3-yl-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-bromo-, monohydrochloride, 5-bromotryptamine hcl, 2-5-bromo-1h-indol-3-yl ethanaminehydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine hydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine-hydrogen chloride 1/1
13241175
2-(5-bromo-1H-indol-3-yl)ethanamine;hydrochloride
C1=CC2=C(C=C1Br)C(=CN2)CCN.Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Bromotryptamine hydrochloride is used as pharmaceutical intermediates.
Notes
Light sensitive. Store in cool dry conditions. Incompatible with light and oxidizing agents. Store in the dark.
TRUSTED_SUSTAINABILITY
Melting Point ∽274°C (decomposition)
Quantity 250 mg
Beilstein 3728224
Sensitivity Light sensitive
Formula Weight 275.58
Percent Purity 98%
Chemical Name or Material 5-Bromotryptamine hydrochloride