missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Bromosalicylic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Bromosalicylic acid is used as organic synthesis. The cross-coupling reaction of 5-bromosalicylic acid with alkyl and aryl Grignard reagents provides a navel efficient synthesis of substituted salicylic acids.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
5-Bromosalicylic acid is used as organic synthesis. The cross-coupling reaction of 5-bromosalicylic acid with alkyl and aryl Grignard reagents provides a navel efficient synthesis of substituted salicylic acids.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 166°C to 170°C |
Quantity | 50 g |
Beilstein | 2209121 |
Formula Weight | 217.02 |
Percent Purity | 98% |
Chemical Name or Material | 5-Bromosalicylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'