Learn More
Lonza™ 5-Bromo-2'Deoxyuridine
Commonly used in the detection of proliferating cells in living tissues.
Brand:  Lonza™ CC-4519
Additional Details : Weight : 1.00000kg
Product Code. 11665530
  • 520.00 NOK / Each
Estimated Shipment: 27-08-2024
Description
  • Synthetic nucleoside that is an analog of thymidine.
  • Commonly used in the detection of proliferating cells in living tissues.
  • Routinely and extensively used to measure DNA synthesis and to label dividing cells.
Specifications
Specifications
Nucleosides
15 mg