Learn More
5-Bromo-1,2,3-trifluorobenzene, 98%, Thermo Scientific Chemicals
Catalog Number p-7025228
Click to view available options
Quantity:
1 g
5 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
138526-69-9
C6H2BrF3
210.981
MFCD00070738
HKJCELUUIFFSIN-UHFFFAOYSA-N
1-bromo-3,4,5-trifluorobenzene, 3,4,5-trifluorobromobenzene, 1,2,3-trifluoro-5-bromobenzene, 5-bromo-1,2,3-trifluoro-benzene, benzene, 5-bromo-1,2,3-trifluoro, pubchem2304, acmc-1c02k, ksc174s3f, 3,4,5-trifluoro-bromobenzene, 3,4,5-trifluorophenyl bromide
611409
5-bromo-1,2,3-trifluorobenzene
C1=C(C=C(C(=C1F)F)F)Br
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
138526-69-9
C6H2BrF3
210.981
MFCD00070738
HKJCELUUIFFSIN-UHFFFAOYSA-N
1-bromo-3,4,5-trifluorobenzene, 3,4,5-trifluorobromobenzene, 1,2,3-trifluoro-5-bromobenzene, 5-bromo-1,2,3-trifluoro-benzene, benzene, 5-bromo-1,2,3-trifluoro, pubchem2304, acmc-1c02k, ksc174s3f, 3,4,5-trifluoro-bromobenzene, 3,4,5-trifluorophenyl bromide
611409
5-bromo-1,2,3-trifluorobenzene
C1=C(C=C(C(=C1F)F)F)Br
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point <-20°C
Density 1.767
Boiling Point 140°C
Flash Point 45°C (113°F)
Refractive Index 1.482
Quantity 1 g
UN Number UN1993
Beilstein 7249191
Formula Weight 210.98
Percent Purity 98%
Chemical Name or Material 5-Bromo-1,2,3-trifluorobenzene