missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Amino-3-methyl-1-phenyl-1H-pyrazole, ≥99%

Brand:  Alfa Aesar™ H63605.14

Additional Details : CAS Number : 1131-18-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15430389

  • 1870.00 NOK / 25g


In Stock 1
Add to basket

Specifications

Chemical Identifiers

1131-18-6
173.219
FMKMKBLHMONXJM-UHFFFAOYSA-N
70801
CC1=NN(C(=C1)N)C2=CC=CC=C2
C10H11N3
MFCD00020727
3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
5-methyl-2-phenylpyrazol-3-amine

Specifications

25g
Brown to Yellow
1131-18-6
MFCD00020727
3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
CC1=NN(C(=C1)N)C2=CC=CC=C2
173.219
173.21
≥99%
5-Amino-3-methyl-1-phenyl-1H-pyrazole
114°C to 117°C
C10H11N3
140666
FMKMKBLHMONXJM-UHFFFAOYSA-N
5-methyl-2-phenylpyrazol-3-amine
70801
Solid
Videos
SDS Documents