Learn More
5-Amino-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™
Catalog Number p-7077421
Click to view available options
Quantity:
1 g
5 g
148890-63-5
C8H8N2O2
164.17
GXGYWLOHPYNWFE-UHFFFAOYSA-N
5-amino-2h-benzo b 1,4 oxazin-3 4h-one, 5-amino-3,4-dihydro-2h-1,4-benzoxazin-3-one, 5-amino-2h-1,4-benzoxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 5-amino, 5-amino-2,4-dihydro-1,4-benzoxazin-3-one, 3,4-dihydro-3-oxo-2h-1,4-benzoxazin-5-amine, 5-amino-4h-benzo 1,4 oxazin-3-one, 5-amino-2h,4h-benzo e 1,4-oxazaperhydroin-3-one
10797099
5-amino-4H-1,4-benzoxazin-3-one
C1C(=O)NC2=C(C=CC=C2O1)N
148890-63-5
C8H8N2O2
164.17
GXGYWLOHPYNWFE-UHFFFAOYSA-N
5-amino-2h-benzo b 1,4 oxazin-3 4h-one, 5-amino-3,4-dihydro-2h-1,4-benzoxazin-3-one, 5-amino-2h-1,4-benzoxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 5-amino, 5-amino-2,4-dihydro-1,4-benzoxazin-3-one, 3,4-dihydro-3-oxo-2h-1,4-benzoxazin-5-amine, 5-amino-4h-benzo 1,4 oxazin-3-one, 5-amino-2h,4h-benzo e 1,4-oxazaperhydroin-3-one
10797099
5-amino-4H-1,4-benzoxazin-3-one
C1C(=O)NC2=C(C=CC=C2O1)N
Melting Point 235.0°C to 241.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.17
Percent Purity 97%
Chemical Name or Material 5-Amino-2H-1,4-benzoxazin-3(4H)-one
compliance-icons
Product Identifier
  • 5-Amino-2H-1,4-benzoxazin-3(4H)-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.