Learn More
5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals
409.22 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 263°C to 265°C |
Quantity | 5 g |
Beilstein | 81983 |
Solubility Information | Soluble in dimethyl sulfoxide. |
Formula Weight | 128.13 |
Percent Purity | ≥98% |
Chemical Name or Material | 5,6-Dihydro-5-methyluracil |