missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Catalog Number p-3522545
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892501-91-6
C10H9BrN2O
253.10
MFCD09064955
GTXIOLDSSMKRDR-UHFFFAOYSA-N
5-2-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 5-2-bromomethyl phenyl-3-methyl-1,2,4-oxadiazol, 1,2,4-oxadiazole, 5-2-bromomethyl phenyl-3-methyl, 2-3-methyl-1,2,4-oxadiazol-5-yl benzyl bromide, pubchem23328, 1,2,4-oxadiazole,5-2-bromomethyl phenyl-3-methyl
24229503
5-[2-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
CC1=NOC(=N1)C1=CC=CC=C1CBr
This item is not returnable. View return policy
892501-91-6
C10H9BrN2O
253.10
MFCD09064955
GTXIOLDSSMKRDR-UHFFFAOYSA-N
5-2-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 5-2-bromomethyl phenyl-3-methyl-1,2,4-oxadiazol, 1,2,4-oxadiazole, 5-2-bromomethyl phenyl-3-methyl, 2-3-methyl-1,2,4-oxadiazol-5-yl benzyl bromide, pubchem23328, 1,2,4-oxadiazole,5-2-bromomethyl phenyl-3-methyl
24229503
5-[2-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
CC1=NOC(=N1)C1=CC=CC=C1CBr
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 5-[2-(Bromomethyl)phenyl]-3-methyl-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'