missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)aniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7046731
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
372-16-7
C7H6F3NS
193.19
MFCD00040926
OHHHTUXVBNGOGI-UHFFFAOYSA-N
4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
123054
NC1=CC=C(SC(F)(F)F)C=C1
This item is not returnable. View return policy
372-16-7
C7H6F3NS
193.19
MFCD00040926
OHHHTUXVBNGOGI-UHFFFAOYSA-N
4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
123054
NC1=CC=C(SC(F)(F)F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.344
Boiling Point 103°C (11 mmHg)
Flash Point 106°C (222°F)
Odor Unpleasant
Refractive Index 1.528
Quantity 1 g
UN Number UN2810
Beilstein 2208859
IUPAC Name 4-(trifluoromethylsulfanyl)aniline
Formula Weight 193.19
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylthio)aniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'