Learn More
4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents.
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents.
Specifications
Melting Point | 60°C to 64°C |
Boiling Point | 119°C to 121°C (17 mmHg) |
Quantity | 1 g |
Beilstein | 743759 |
Solubility Information | Insoluble in water. |
Formula Weight | 176.14 |
Percent Purity | 95% |
Chemical Name or Material | 4-(Trifluoromethyl)phenylhydrazine |