missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzaldehyde, 97%, Thermo Scientific Chemicals
375.61 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.
Solubility
Soluble in water.
Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.
Solubility
Soluble in water.
Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications
Density | 0.97 |
Boiling Point | 244°C to 246°C |
Flash Point | 101°C (213°F) |
Refractive Index | 1.5285 |
Quantity | 5 g |
UN Number | UN3082 |
Beilstein | 1906461 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 162.23 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'