Learn More
4-Nitrophenyl-beta-D-glucuronide, 99+%, Thermo Scientific Chemicals
Catalog Number p-174528
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
10344-94-2
C12H12NO9
314.23
MFCD00036210
QSUILVWOWLUOEU-GOVZDWNOSA-M
4-nitrophenyl-beta-d-glucuronide, 4-nitrophenylglucuronide, 4-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl beta-d-glucuronide, p-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl b-d-glucosiduronic acid, p-nitrophenyl-b-d-glucuronide, p-nitrophenyl beta-d-glucuronide, beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14, 4-nitrophenyl-beta-d-glucopyranosiduronic acid
82565
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O
10344-94-2
C12H12NO9
314.23
MFCD00036210
QSUILVWOWLUOEU-GOVZDWNOSA-M
4-nitrophenyl-beta-d-glucuronide, 4-nitrophenylglucuronide, 4-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl beta-d-glucuronide, p-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl b-d-glucosiduronic acid, p-nitrophenyl-b-d-glucuronide, p-nitrophenyl beta-d-glucuronide, beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14, 4-nitrophenyl-beta-d-glucopyranosiduronic acid
82565
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.05% max. (p-Nitrophenol) (HPLC)
Packaging Glass bottle
Quantity 50 mg
Specific Rotation −107° to −113° (20°C, 589nm) (c=1, H2O)
Solubility Information (1% in water) Clear colorless to light yellow
Formula Weight 315.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Nitrophenyl-β-D-glucuronide