missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octylphenol, 99%, Thermo Scientific Chemicals
266.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 43°C to 45°C |
Density | 0.961 |
Boiling Point | 150°C (4 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 1 g |
UN Number | UN2430 |
Solubility Information | Insoluble in water. |
Formula Weight | 206.32 |
Percent Purity | 99% |
Chemical Name or Material | 4-n-Octylphenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'