Learn More
4-(Methylthio)phenol, 98%, Thermo Scientific Chemicals
Catalog Number p-7028816
Click to view available options
Quantity:
10 g
50 g
250 g
1073-72-9
C7H8OS
140.20
MFCD00002351
QASBCTGZKABPKX-UHFFFAOYSA-N
4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide
14086
CHEBI:38862
4-methylsulfanylphenol
CSC1=CC=C(O)C=C1
1073-72-9
C7H8OS
140.20
MFCD00002351
QASBCTGZKABPKX-UHFFFAOYSA-N
4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide
14086
CHEBI:38862
4-methylsulfanylphenol
CSC1=CC=C(O)C=C1
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water 9.59 g/ l.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 85°C to 87°C
Boiling Point 153°C to 156°C (20 mmHg)
Odor Stench
Quantity 10 g
Solubility Information Soluble in water 9.59g/ l.
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)phenol