missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylsalicylic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.
Solubility
Soluble in water 10 g/L @ 20˚C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.
Solubility
Soluble in water 10 g/L @ 20˚C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 176°C to 180°C |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 2208140 |
Merck Index | 14,2580 |
Solubility Information | Soluble in water 10g/L @ 20?C. |
Formula Weight | 152.15 |
Percent Purity | 99% |
Chemical Name or Material | 4-Methylsalicylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'