Learn More
4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid, 95%, Thermo Scientific™
Catalog Number 15269726
Click to view available options
Quantity:
1 g
5 g
1332627-30-1
C11H13NO2
191.23
QOQHRSQQPZYCGY-UHFFFAOYSA-N
1,2,3,4-tetrahydro-4-methylquinoline-6-carboxylic acid
56766060
4-methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
CC1CCNC2=C1C=C(C=C2)C(=O)O
1332627-30-1
C11H13NO2
191.23
QOQHRSQQPZYCGY-UHFFFAOYSA-N
1,2,3,4-tetrahydro-4-methylquinoline-6-carboxylic acid
56766060
4-methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
CC1CCNC2=C1C=C(C=C2)C(=O)O
Melting Point 158°C to 160°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 191.23
Percent Purity 95%
Chemical Name or Material 4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
compliance-icons
Product Identifier
  • 4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.