Learn More
Alfa Aesar™ 4-Methoxyindole, 99%
Brand: Alfa Aesar™ L19069.MD
Additional Details : CAS Number : 4837-90-5 Weight : 0.01000kg
Quantity | 250mg |
---|
Chemical Identifiers
4837-90-5 | |
147.177 | |
LUNOXNMCFPFPMO-UHFFFAOYSA-N | |
138363 | |
COC1=CC=CC2=C1C=CN2 |
C9H9NO | |
MFCD00009737 | |
4-methoxyindole, 1h-indole, 4-methoxy, 4-methoxylindole, 1h-indol-4-yl methyl ether, indole, 4-methoxy, 4-methoxy-indole, zlchem 710, pubchem7433, acmc-1aoxe | |
4-methoxy-1H-indole |
Specifications
4-Methoxyindole | |
181°C to 183°C (24mmHg) | |
250mg | |
C9H9NO | |
122456 | |
LUNOXNMCFPFPMO-UHFFFAOYSA-N | |
4-methoxy-1H-indole | |
138363 | |
99% |
Light sensitive | |
69°C to 71°C | |
4837-90-5 | |
MFCD00009737 | |
4-methoxyindole, 1h-indole, 4-methoxy, 4-methoxylindole, 1h-indol-4-yl methyl ether, indole, 4-methoxy, 4-methoxy-indole, zlchem 710, pubchem7433, acmc-1aoxe | |
COC1=CC=CC2=C1C=CN2 | |
147.177 | |
147.18 |