missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester is used as organic reagents.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester is used as organic reagents.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 48°C to 51°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 296.56 |
Percent Purity | 98% |
Chemical Name or Material | 4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'